2-Amino-2-methyl-1-(1-pyrrolidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052332-500MG 052332-2.5G
101899-214EA 364.62 USD
101899-214 101899-216
2-Amino-2-methyl-1-(1-pyrrolidinyl)-1-propanone hydrochloride
2-Amino-2-methyl-1-(1-pyrrolidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD13562092
CAS Number: 1220031-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR