2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052328-500MG 052328-2.5G
101899-198EA 364.62 USD
101899-198 101899-200
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-3-methyl-1-butanone hydrochloride
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O
MDL Number: MFCD13562088
CAS Number: 1246172-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR