2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052327-500MG 052327-2.5G
101899-194EA 364.62 USD
101899-194 101899-196
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-3-methyl-1-butanone hydrochloride
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562087
CAS Number: 1236259-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR