2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052325-500MG 052325-2.5G
101899-186EA 364.62 USD
101899-186 101899-188
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-3-methyl-1-butanone hydrochloride
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄ClN₃O₂
MW: 265.79 g/mol
MDL Number: MFCD13562085
CAS Number: 1236259-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR