2-Amino-N-cyclohexyl-N-ethyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052324-500MG 052324-2.5G
101899-182EA 364.62 USD
101899-182 101899-184
2-Amino-N-cyclohexyl-N-ethyl-3-methylbutanamide hydrochloride
2-Amino-N-cyclohexyl-N-ethyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₇ClN₂O
MDL Number: MFCD13562084
CAS Number: 1236254-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR