2-Amino-N-benzyl-N,3-dimethylbutanamide hydrochloride

Supplier: Matrix Scientific

052323-500MG 052323-2.5G
101899-178EA 364.62 USD
101899-178 101899-180
2-Amino-N-benzyl-N,3-dimethylbutanamide hydrochloride
2-Amino-N-benzyl-N,3-dimethylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O
MDL Number: MFCD13562083
CAS Number: 1236262-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR