2-Amino-1-(2-ethyl-1-piperidinyl)-3-methyl-1-butanone hydrochloride
Supplier: Matrix Scientific
052322-500MG
052322-2.5G
101899-174EA
364.62
USD
101899-174
101899-176
2-Amino-1-(2-ethyl-1-piperidinyl)-3-methyl-1-butanone hydrochloride
2-Amino-1-(2-ethyl-1-piperidinyl)-3-methyl-1-butanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₅ClN₂O |
MDL Number:
MFCD13562082 CAS Number: 1236266-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...