2-Amino-N-cyclohexyl-N-3-dimethylbutanamide hydrochloride

Supplier: Matrix Scientific

052321-500MG 052321-2.5G
101899-170EA 364.62 USD
101899-170 101899-172
2-Amino-N-cyclohexyl-N-3-dimethylbutanamide hydrochloride
2-Amino-N-cyclohexyl-N-3-dimethylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₅ClN₂O
MDL Number: MFCD13562081
CAS Number: 1236258-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR