2-Amino-3-methyl-N,N-dipropylbutanamide hydrochloride

Supplier: Matrix Scientific

052320-500MG 052320-2.5G
101899-166EA 364.62 USD
101899-166 101899-168
2-Amino-3-methyl-N,N-dipropylbutanamide hydrochloride
2-Amino-3-methyl-N,N-dipropylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅ClN₂O
MW: 236.79 g/mol
MDL Number: MFCD13562080
CAS Number: 1236260-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR