2-Amino-1-(4-benzyl-1-piperazinyl)-3-methyl-1-butanone hydrochloride

Supplier: Matrix Scientific

052316-500MG 052316-2.5G
101899-150EA 364.62 USD
101899-150 101899-152
2-Amino-1-(4-benzyl-1-piperazinyl)-3-methyl-1-butanone hydrochloride
2-Amino-1-(4-benzyl-1-piperazinyl)-3-methyl-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClN₃O
MDL Number: MFCD13562076
CAS Number: 1236259-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR