N,N-Diallyl-2-amino-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052311-500MG 052311-2.5G
101899-130EA 364.62 USD
101899-130 101899-132
N,N-Diallyl-2-amino-3-methylbutanamide hydrochloride
N,N-Diallyl-2-amino-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O
MDL Number: MFCD13562071
CAS Number: 1246172-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR