2-Amino-N-ethyl-N-(2-hydroxyethyl)-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052310-500MG 052310-2.5G
101899-126EA 364.62 USD
101899-126 101899-128
2-Amino-N-ethyl-N-(2-hydroxyethyl)-3-methylbutanamide hydrochloride
2-Amino-N-ethyl-N-(2-hydroxyethyl)-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₉H₂₁ClN₂O₂
MW: 224.73 g/mol
MDL Number: MFCD13562070
CAS Number: 1236256-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR