2-Amino-N-butyl-N-3-dimethylbutanamide hydrochloride
Supplier: Matrix Scientific
052309-500MG
052309-2.5G
101899-122EA
364.62
USD
101899-122
101899-124
2-Amino-N-butyl-N-3-dimethylbutanamide hydrochloride
2-Amino-N-butyl-N-3-dimethylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₃ClN₂O |
MDL Number:
MFCD13562069 CAS Number: 1236258-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...