2-Amino-3-methyl-1-(4-morpholinyl)-1-butanone hydrochloride

Supplier: Matrix Scientific

052307-500MG 052307-2.5G
101899-114EA 166.15 USD
101899-114 101899-116
2-Amino-3-methyl-1-(4-morpholinyl)-1-butanone hydrochloride
2-Amino-3-methyl-1-(4-morpholinyl)-1-butanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562067
CAS Number: 56415-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR