2-Amino-3-methyl-1-(1-piperidinyl)-1-butanone hydrochloride

Supplier: Matrix Scientific

052306-500MG 052306-2.5G
101899-110EA 364.62 USD
101899-110 101899-112
2-Amino-3-methyl-1-(1-piperidinyl)-1-butanone hydrochloride
2-Amino-3-methyl-1-(1-piperidinyl)-1-butanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562066
CAS Number: 56415-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR