2-Amino-N-(2-hydroxyethyl)-N,3-dimethylbutanamide hydrochloride
Supplier: Matrix Scientific
052305-500MG
052305-2.5G
101899-106EA
364.62
USD
101899-106
101899-108
2-Amino-N-(2-hydroxyethyl)-N,3-dimethylbutanamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-N,3-dimethylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₉ClN₂O₂ |
MDL Number:
MFCD13562065 CAS Number: 1236255-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...