2-Amino-3-methyl-1-(1-pyrrolidinyl)-1-butanone hydrochloride

Supplier: Matrix Scientific

052303-500MG 052303-2.5G
101899-098EA 364.62 USD
101899-098 101899-100
2-Amino-3-methyl-1-(1-pyrrolidinyl)-1-butanone hydrochloride
2-Amino-3-methyl-1-(1-pyrrolidinyl)-1-butanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562063
CAS Number: 1236266-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR