2-Amino-N,N-3-trimethylbutanamide hydrochloride

Supplier: Matrix Scientific

052302-500MG 052302-2.5G
101899-094EA 364.62 USD
101899-094 101899-096
2-Amino-N,N-3-trimethylbutanamide hydrochloride
2-Amino-N,N-3-trimethylbutanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13562062
CAS Number: 1257848-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR