3-Amino-1-(2-ethyl-1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052293-500MG 052293-2.5G
101899-058EA 364.62 USD
101899-058 101899-060
3-Amino-1-(2-ethyl-1-piperidinyl)-1-propanone hydrochloride
3-Amino-1-(2-ethyl-1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562054
CAS Number: 1220033-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR