3-Amino-N-cyclohexyl-N-methylpropanamide hydrochloride
Supplier: Matrix Scientific
052292-500MG
052292-2.5G
101899-054EA
364.62
USD
101899-054
101899-056
3-Amino-N-cyclohexyl-N-methylpropanamide hydrochloride
3-Amino-N-cyclohexyl-N-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O |
MDL Number:
MFCD13562053 CAS Number: 1219971-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...