4-(3-Aminopropanoyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052289-500MG 052289-2.5G
101899-042EA 364.62 USD
101899-042 101899-044
4-(3-Aminopropanoyl)-2-piperazinone hydrochloride
4-(3-Aminopropanoyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClN₃O₂
MDL Number: MFCD13562050
CAS Number: 1220033-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR