3-Amino-1-(1-azepanyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052288-500MG 052288-2.5G
101899-038EA 364.62 USD
101899-038 101899-040
3-Amino-1-(1-azepanyl)-1-propanone hydrochloride
3-Amino-1-(1-azepanyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562049
CAS Number: 1220039-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR