3-Amino-1-(4-benzyl-1-piperazinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052287-500MG 052287-2.5G
101899-034EA 364.62 USD
101899-034 101899-036
3-Amino-1-(4-benzyl-1-piperazinyl)-1-propanone hydrochloride
3-Amino-1-(4-benzyl-1-piperazinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClN₃O
MW: 283.8 g/mol
MDL Number: MFCD13562048
CAS Number: 1220031-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR