3-Amino-1-(3-methyl-1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052284-500MG 052284-2.5G
101899-022EA 364.62 USD
101899-022 101899-024
3-Amino-1-(3-methyl-1-piperidinyl)-1-propanone hydrochloride
3-Amino-1-(3-methyl-1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562045
CAS Number: 1220034-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR