3-Amino-1-(2-methyl-1-piperidinyl)-1-propanone hydrochloride
Supplier: Matrix Scientific
052283-500MG
052283-2.5G
101899-018EA
364.62
USD
101899-018
101899-020
3-Amino-1-(2-methyl-1-piperidinyl)-1-propanone hydrochloride
3-Amino-1-(2-methyl-1-piperidinyl)-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O |
MDL Number:
MFCD13562044 CAS Number: 1220017-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...