3-Amino-N-ethyl-N-(2-hydroxyethyl)propanamide hydrochloride

Supplier: Matrix Scientific

052281-500MG 052281-2.5G
101899-010EA 364.62 USD
101899-010 101899-012
3-Amino-N-ethyl-N-(2-hydroxyethyl)propanamide hydrochloride
3-Amino-N-ethyl-N-(2-hydroxyethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562042
CAS Number: 1220031-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR