3-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052279-500MG 052279-2.5G
101899-002EA 364.62 USD
101899-002 101899-004
3-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-propanone hydrochloride
3-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O₂
MDL Number: MFCD13562040
CAS Number: 1220038-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR