3-Amino-1-(4-morpholinyl)-1-propanone hydrochloride
Supplier: Matrix Scientific
052278-500MG
052278-2.5G
101898-998EA
86.15
USD
101898-998
101899-000
3-Amino-1-(4-morpholinyl)-1-propanone hydrochloride
3-Amino-1-(4-morpholinyl)-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅ClN₂O₂ |
MDL Number:
MFCD07366749 CAS Number: 173336-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...