3-Amino-N-(2-hydroxyethyl)-N-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052277-500MG 052277-2.5G
101898-994EA 186.15 USD
101898-994 101898-996
3-Amino-N-(2-hydroxyethyl)-N-methylpropanamide hydrochloride
3-Amino-N-(2-hydroxyethyl)-N-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13562039
CAS Number: 1220033-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR