3-Amino-N,N-diethylpropanamide hydrochloride

Supplier: Matrix Scientific

052276-500MG 052276-2.5G
101898-990EA 364.62 USD
101898-990 101898-992
3-Amino-N,N-diethylpropanamide hydrochloride
3-Amino-N,N-diethylpropanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD07366747
CAS Number: 34105-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR