3-Amino-N,N-dimethylpropanamide hydrochloride

Supplier: Matrix Scientific

052274-500MG 052274-2.5G
101898-982EA 280 USD
101898-982 101898-984
3-Amino-N,N-dimethylpropanamide hydrochloride
3-Amino-N,N-dimethylpropanamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O
MDL Number: MFCD10686752
CAS Number: 1857-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR