N-Benzyl-N-ethyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052272-500MG 052272-2.5G
101898-974EA 364.62 USD
101898-974 101898-976
N-Benzyl-N-ethyl-2-(methylamino)acetamide hydrochloride
N-Benzyl-N-ethyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD13562037
CAS Number: 1219972-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR