1-[3,4-Dihydro-1(2H)-quinolinyl]-2-(methylamino)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052271-500MG 052271-2.5G
101898-970EA 364.62 USD
101898-970 101898-972
1-[3,4-Dihydro-1(2H)-quinolinyl]-2-(methylamino)-1-ethanone hydrochloride
1-[3,4-Dihydro-1(2H)-quinolinyl]-2-(methylamino)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD13562036
CAS Number: 1220036-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR