1-(2,3-Dihydro-1H-indol-1-yl)-2-(methylamino)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052270-500MG 052270-2.5G
101898-966EA 384.62 USD
101898-966 101898-968
1-(2,3-Dihydro-1H-indol-1-yl)-2-(methylamino)-1-ethanone hydrochloride
1-(2,3-Dihydro-1H-indol-1-yl)-2-(methylamino)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD07286204
CAS Number: 1049769-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR