1-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-(methylamino)-1-ethanone hydrochloride
Supplier: Matrix Scientific
052268-500MG
052268-2.5G
101898-958EA
364.62
USD
101898-958
101898-960
1-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-(methylamino)-1-ethanone hydrochloride
1-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-(methylamino)-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₉H₂₀ClN₃O₂ |
MDL Number:
MFCD13562034 CAS Number: 1220017-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...