N-Benzyl-N-methyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052266-500MG 052266-2.5G
101898-950EA 364.62 USD
101898-950 101898-952
N-Benzyl-N-methyl-2-(methylamino)acetamide hydrochloride
N-Benzyl-N-methyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O
MDL Number: MFCD13562032
CAS Number: 1220033-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR