1-(2-Ethyl-1-piperidinyl)-2-(methylamino)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052265-500MG 052265-2.5G
101898-946EA 364.62 USD
101898-946 101898-948
1-(2-Ethyl-1-piperidinyl)-2-(methylamino)-1-ethanone hydrochloride
1-(2-Ethyl-1-piperidinyl)-2-(methylamino)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562031
CAS Number: 1220033-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR