1-(4-Hydroxy-1-piperidinyl)-2-(methylamino)-1-ethanone hydrochloride
Supplier: Matrix Scientific
052262-500MG
052262-2.5G
101898-934EA
364.62
USD
101898-934
101898-936
1-(4-Hydroxy-1-piperidinyl)-2-(methylamino)-1-ethanone hydrochloride
1-(4-Hydroxy-1-piperidinyl)-2-(methylamino)-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ |
MDL Number:
MFCD12913018 CAS Number: 1181868-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...