4-[2-(Methylamino)acetyl]-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052261-500MG 052261-2.5G
101898-930EA 364.62 USD
101898-930 101898-932
4-[2-(Methylamino)acetyl]-2-piperazinone hydrochloride
4-[2-(Methylamino)acetyl]-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClN₃O₂
MDL Number: MFCD13562028
CAS Number: 1220017-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR