1-(1-Azepanyl)-2-(methylamino)-1-ethanone hydrochloride
Supplier: Matrix Scientific
052260-500MG
052260-2.5G
101898-926EA
364.62
USD
101898-926
101898-928
1-(1-Azepanyl)-2-(methylamino)-1-ethanone hydrochloride
1-(1-Azepanyl)-2-(methylamino)-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O |
MDL Number:
MFCD13562027 CAS Number: 1220033-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...