2-(Methylamino)-1-(4-methyl-1-piperazinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052258-500MG 052258-2.5G
101898-918EA 364.62 USD
101898-918 101898-920
2-(Methylamino)-1-(4-methyl-1-piperazinyl)-1-ethanone hydrochloride
2-(Methylamino)-1-(4-methyl-1-piperazinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClN₃O
MDL Number: MFCD13562026
CAS Number: 1219968-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR