2-(Methylamino)-1-(2-methyl-1-piperidinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052255-500MG 052255-2.5G
101898-906EA 364.62 USD
101898-906 101898-908
2-(Methylamino)-1-(2-methyl-1-piperidinyl)-1-ethanone hydrochloride
2-(Methylamino)-1-(2-methyl-1-piperidinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562023
CAS Number: 1220038-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR