N,N-Diallyl-2-(methylamino)acetamide hydrochloride
Supplier: Matrix Scientific
052254-500MG
052254-2.5G
101898-902EA
364.62
USD
101898-902
101898-904
N,N-Diallyl-2-(methylamino)acetamide hydrochloride
N,N-Diallyl-2-(methylamino)acetamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₇ClN₂O |
MDL Number:
MFCD13562022 CAS Number: 1219972-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...