N-Ethyl-N-(2-hydroxyethyl)-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052253-500MG 052253-2.5G
101898-898EA 364.62 USD
101898-898 101898-900
N-Ethyl-N-(2-hydroxyethyl)-2-(methylamino)acetamide hydrochloride
N-Ethyl-N-(2-hydroxyethyl)-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562021
CAS Number: 1219982-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR