N-Butyl-N-methyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052252-500MG 052252-2.5G
101898-894EA 364.62 USD
101898-894 101898-896
N-Butyl-N-methyl-2-(methylamino)acetamide hydrochloride
N-Butyl-N-methyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O
MDL Number: MFCD13562020
CAS Number: 1220036-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR