1-(3-Hydroxy-1-pyrrolidinyl)-2-(methylamino)-1-ethanone hydrochloride
Supplier: Matrix Scientific
052251-500MG
052251-2.5G
101898-890EA
364.62
USD
101898-890
101898-892
1-(3-Hydroxy-1-pyrrolidinyl)-2-(methylamino)-1-ethanone hydrochloride
1-(3-Hydroxy-1-pyrrolidinyl)-2-(methylamino)-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅ClN₂O₂ |
MDL Number:
MFCD13562019 CAS Number: 1219972-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...