2-(Methylamino)-1-(4-morpholinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052250-500MG 052250-2.5G
101898-886EA 364.62 USD
101898-886 101898-888
2-(Methylamino)-1-(4-morpholinyl)-1-ethanone hydrochloride
2-(Methylamino)-1-(4-morpholinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O₂
MDL Number: MFCD08060102
CAS Number: 41458-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR