N-(2-Hydroxyethyl)-N-methyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052248-500MG 052248-2.5G
101898-878EA 364.62 USD
101898-878 101898-880
N-(2-Hydroxyethyl)-N-methyl-2-(methylamino)acetamide hydrochloride
N-(2-Hydroxyethyl)-N-methyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13562018
CAS Number: 1219982-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR