N,N-Diethyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052247-500MG 052247-2.5G
101898-874EA 364.62 USD
101898-874 101898-876
N,N-Diethyl-2-(methylamino)acetamide hydrochloride
N,N-Diethyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MW: 180.68 g/mol
MDL Number: MFCD07308365
CAS Number: 877880-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR