2-(Methylamino)-1-(1-pyrrolidinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052246-500MG 052246-2.5G
101898-870EA 364.62 USD
101898-870 101898-872
2-(Methylamino)-1-(1-pyrrolidinyl)-1-ethanone hydrochloride
2-(Methylamino)-1-(1-pyrrolidinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD08060488
CAS Number: 144685-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR